Learn More
Thermo Scientific™ Nunc™ Press Out Tool for Cryobank and Bank-It™ Vial Systems
Minimize freeze/thaw by using the Thermo Scienctific™ Nunc™ Press Out Tool for Cryobank and Bank-It™ Systems, which allows for rapid retrieval of Cryobank vials.
Brand: Thermo Scientific™ 374009
996.00 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : 76
Additional Details : Weight : 1.10000kg
Description
Locate and remove a desired tube quickly and easily, without allowing frozen samples to thaw by using the Thermo Scientific™ Nunc™ Press Out Tool for Cryobank and Bank-It™ Vial Systems.
Features:- Selected vials are released by levers and can be easily harvested
- Selected position can be chosen based on data base output
- Pegs can be placed and checked while the sample/neighboring samples remain cold
- Cut-off corner ensures the rack is orientated correctly
Specifications
Nunc Cryobank and Bank-it Rack, Tube Press out Tool, 1 Each | |
Cryobank | |
1 | |
Cryobank and Bank-It Systems |
Tube Press Out Tool | |
Silver | |
1 |