missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Rectangular Dishes

Product Code. p-4524632
Click to view available options
No. of Wells:
1
4
8
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 11722515

Brand: Thermo Scientific™ 267060

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Perform molecular biology applications with non-treated Thermo Scientific™ Nunc™ Rectangular Dishes, which have ANSI™ standard dimensions and are optimized for robotics.

Non-treated Thermo Scientific™ Nunc™ Rectangular Dishes with ANSI™ standard dimensions are optimized for molecular biology applications using robotic instruments.

Rectangular Dishes 1, 4 and 8-Well

  • 1-well dish is ideal for HTS Petri dish applications
  • 4- and 8-well dishes are ideal for assay compartmentalization, probe multiple gene sub-set
  • 4-well dish holds four glass slides for storage

1-Well Format

  • Convenient plate format for cell culture
  • HTS Petri dish applications

4- and 8-Well Formats

  • Assay compartmentalization
  • Probe multiple gene subsets
  • Wash Western blot strips
  • 4-well dish holds four glass slides
  • Useful in all areas of cell culture, including scale-up and cloning

Ordering Information:

Customized bar coding is available

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Product Type Cell Culture Dish
No. of Wells 1
Sterility Sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 90 mL
Well Shape Rectangular
Includes Lid
No. per Case 100
No. per Pack 10
Length (Metric) 100 mm
Width (Metric) 86 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.