Learn More
Thermo Scientific™ Nunc™ Screwdrivers for Cryobank and Bank-It™ Vial Systems, manual
Quickly and easily cap and decap Thermo Scientific™ Nunc™ Cryobank tubes using these screwdrivers, available in manual and electric models to suit your application.
£45.90 - £3875.00
Specifications
Coding | 2D Barcode |
---|---|
Vial Type | Cryobank, Bank-It |
For Use With (Equipment) | Cryobank and Bank-It Systems |
Product Code | Brand | Description | Plug Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Plug Type | Price | Quantity & Availability | ||||
10709153
|
Thermo Scientific™
369643 |
Manual Screwdriver | US |
£45.90
Each |
In Stock
Log in to see stock. |
||||
10401633
|
Thermo Scientific™
369644 |
Electric Screwdriver, European Plug | European | N/A | |||||
Description
Quickly and easily cap and decap Nunc Cryobank tubes using Thermo Scientific™ Nunc™ Screwdrivers for Cryobank and Bank-It™ Systems, available in manual and electric models to suit your application. Nunc Cryobank vials provide the customer with superior tracking of their cryovials using a 2D code, the highest storage density on the market and the ability to automate the capping and decapping of the vials. The tubes click into the rack and can be handled without the risk of accidentally dropping a vial.
Screwdriver socket retains the vial cap, allowing one-handed manipulation of the cap.Specifications
2D Barcode | |
Cryobank, Bank-It | |
Cryobank and Bank-It Systems |