missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ Seals, Tapes, and Foils

Click to view available options
Material:
Aluminum
Non-woven Rayon
PVC-coated Polypropylene
Plastic
Polyester
Polyethylene
Polyolefin
Polypropylene
Vinyl
For Use With (Application):
Breathable, Cell culture
Cell culture, sterile applications
Cell growth, Bacterial growth
Consistent seal
Crystallography and genotyping
Fluorescence, photosensitive samples
General purpose, ELISA
General purpose, sample detection
Low autofluorescence, ELISA
Luminescence, photosensitive samples
PCR
PCR, fluorescence assays, microscopy
PCR, long-term storage
PCR, qPCR, ELISA, Protein crystallography
PCR, storage in 384 well plates
PCR, storage in 96-well plates
Short-term storage, ELISA
Short-term storage, ELISA, minimizes evaporation and cross contamination
Ventilating and storing cell cultures
Certified RNase/DNase-free:
No
Yes
Unit Size:
Pack of 100
Pack of 1000
Pack of 2
Pack of 200
Pack of 50
Pack of 800
Description
Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.
- Adhesive seals effectively seal all microplate formats
- Pressure sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
- Thin, lightweight tapes seal onto the plate with a convenient, handheld applicator (available separately, Cat. No. 250050)
Specifications
Specifications
| Color | White |
| Solvent Or Alcohol Resistant | Life Science Reagent Resistant |
| Description | SEALING TAPE WHITE RAYON |
| Material | Non-woven Rayon |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| No. of Wells | 96 |
| No. per Case | 50 |
| No. per Pack | 50 |
| For Use With (Application) | Cell growth, Bacterial growth |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction