missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ Seals, Tapes, and Foils
Minimize evaporation and protect samples from contamination and spilling with these sealing tapes, ideal for all assay applications.
Brand: Thermo Scientific™ 236366
345.53 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Description
Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.
- Adhesive seals effectively seal all microplate formats
- Pressure sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
- Thin, lightweight tapes seal onto the plate with a convenient, handheld applicator (available separately, Cat. No. 250050)
Specifications
Clear | |
SEALING TAPE POLYESTR STRL CLR | |
Sterile | |
96 | |
200 | |
General | |
Nunc | |
No | |
-40°C to +120°C | |
All Microplates | |
Yes |
Life Science Reagent Resistant | |
Polyester | |
Not Autoclavable | |
800 | |
Cell culture; sterile applications; ELISA | |
Adhesive Seal | |
Acrylic | |
No | |
All well shapes | |
General |