missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Seals, Tapes, and Foils
GREENER_CHOICE

Product Code. 10194411
Click to view available options
Material:
Aluminum
Non-woven Rayon
PVC-coated Polypropylene
Plastic
Polyester
Polyethylene
Polyolefin
Polypropylene
Vinyl
For Use With (Application):
Breathable, Cell culture
Cell culture, sterile applications
Cell growth, Bacterial growth
Consistent seal
Crystallography and genotyping
Fluorescence, photosensitive samples
General purpose, ELISA
General purpose, sample detection
Low autofluorescence, ELISA
Luminescence, photosensitive samples
PCR
PCR, fluorescence assays, microscopy
PCR, long-term storage
PCR, qPCR, ELISA, Protein crystallography
PCR, storage in 384 well plates
PCR, storage in 96-well plates
Short-term storage, ELISA
Short-term storage, ELISA, minimizes evaporation and cross contamination
Ventilating and storing cell cultures
Certified RNase/DNase-free:
No
Yes
Unit Size:
Pack of 100
Pack of 1000
Pack of 2
Pack of 200
Pack of 50
Pack of 800
This item is not returnable. View return policy

Product Code. 10194411

Brand: Thermo Scientific™ 232702

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Minimize evaporation and protect samples from contamination and spilling with these sealing tapes, ideal for all assay applications.

Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.

  • Adhesive seals effectively seal all microplate formats
  • Pressure sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
  • Thin, lightweight tapes seal onto the plate with a convenient, handheld applicator (available separately, Cat. No. 250050)
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Solvent Or Alcohol Resistant Life Science Reagent Resistant
Description AMPLIFICATION TAPE
Material Polypropylene
Sterility Non-sterile
Autoclavable Not Autoclavable
No. of Wells 96
No. per Case 100
No. per Pack 100
For Use With (Application) PCR, fluorescence assays, microscopy
Content And Storage General; Long term
Product Type Adhesive Seal
Product Line Nunc
Adhesive Acrylic
Certified RNase/DNase-free Yes
Pierceable No
Temperature Range (Metric) -70°C to +100°C
Well Shape All well shapes
Plate Compatibility All Microplates
Incubation Conditions General
Peelable Yes
Fluorescence Low Autofluorescence
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.