missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ Seals, Tapes, and Foils
Minimize evaporation and protect samples from contamination and spilling with these sealing tapes, ideal for all assay applications.
Brand: Thermo Scientific™ 241205
185.75 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Description
Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.
- Adhesive seals effectively seal all microplate formats
- Pressure sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
- Thin, lightweight tapes seal onto the plate with a convenient, handheld applicator (available separately, Cat. No. 250050)
Specifications
White | |
Sealing Tape Rayon White Ster | |
Sterile | |
96 | |
25 | |
Adhesive Seal | |
Acrylic | |
Yes | |
All well shapes | |
General | |
Yes |
Life Science Reagent Resistant | |
Non-woven Rayon | |
Not Autoclavable | |
50 | |
Cell growth, Bacterial growth, Non cytotoxic, Moisture vapor transmission rate of 4.200 g/m^2/24 hrs, highly gas permeable | |
Nunc | |
No | |
-20°C to +80°C | |
All Microplates | |
Yes |