Learn More
Thermo Scientific™ Nunc™ Seals, Tapes, and Foils
Minimize evaporation and protect samples from contamination and spilling with these sealing tapes, ideal for all assay applications.
Brand: Thermo Scientific™ 232699
74.37 GBP valid until 2024-07-31
Use promo code "21836" to get your promotional price.
Code : 76
Additional Details : Weight : 0.35000kg
Description
Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.
- Adhesive seals effectively seal all microplate formats
- Pressure sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
- Thin, lightweight tapes seal onto the plate with a convenient, handheld applicator (available separately, Cat. No. 250050)
Specifications
Foil | |
AL SEAL TAPE FOR 384 WELL PLT | |
Non-sterile | |
384 | |
100 | |
PCR and storage in 384 well plates | |
Adhesive Seal | |
Acrylic | |
Yes | |
All well shapes | |
General |
Life Science Reagent Resistant | |
Aluminum | |
Not Autoclavable | |
100 | |
Malleable allows for visual verification of seal security, minimizes evaporation and cross contamination | |
General; Long term | |
Nunc | |
Yes | |
-80°C to +120°C | |
All Microplates | |
Yes |