Learn More
Thermo Scientific™ Nunc™ Seals, Tapes, and Foils

Minimize evaporation and protect samples from contamination and spilling with these sealing tapes, ideal for all assay applications.
Brand: Thermo Scientific™ 232698
54.08 GBP valid until 2025-06-30
Use promo code "25075" to get your promotional price.
Description
Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.
- Adhesive seals effectively seal all microplate formats
- Pressure sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
- Thin, lightweight tapes seal onto the plate with a convenient, handheld applicator (available separately, Cat. No. 250050)

Specifications
Foil | |
AL SEAL TAPE FOR 96 WELL PLT | |
Non-sterile | |
96 | |
100 | |
PCR, storage in 96-well plates | |
Adhesive Seal | |
Acrylic | |
Yes | |
All well shapes | |
General |
Life Science Reagent Resistant | |
Aluminum | |
Not Autoclavable | |
100 | |
Malleable allows for visual verification of seal security, minimizes evaporation and cross contamination | |
General; Long term | |
Nunc | |
Yes | |
-80°C to +120°C | |
All Microplates | |
Yes |
Special Offers
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
-
Thermo Scientific Matrix
Exclusive Discounts on Storage Solutions: Storage Tubes, Equipment and Accessories
Upgrade your lab with top-quality storage consumables and equipment at unbeatable prices.Promo Code: 25075Expires: 2025-06-30
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.