Learn More
Thermo Scientific™ Nunc™ Seals, Tapes, and Foils
Minimize evaporation and protect samples from contamination and spilling with these sealing tapes, ideal for all assay applications.
Brand: Thermo Scientific™ AB-0580
36.09 GBP valid until 2024-07-31
Use promo code "21836" to get your promotional price.
Code : 03
Additional Details : Weight : 0.03960kg
Description
Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.
Thermo Scientific Adhesive Plate Seals provide an effective seal. They feature an easy-removal non-adhesive tab and can withstand temperatures between -40°C to 80°C.Features of Thermo Scientific Adhesive Plate Seals
- Non-adhesive tab for easy removal of seal
- Broad temperature range: -40°C to 80°C
Specifications
Clear | |
Adhesive Plate Seals | |
Non-sterile | |
96 | |
100 | |
General; For long or short term storage | |
Abgene | |
No | |
-40°C to +80°C | |
All Microplates | |
Yes |
Life Science Reagent Resistant | |
PVC-coated Polypropylene | |
Autoclavable | |
100 | |
For crystallography and genotyping | |
Adhesive Seal | |
Acrylic | |
Yes | |
All well shapes | |
General |