missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Square BioAssay Dishes
BIOPHARMA

Product Code. p-4524665
Click to view available options
Height (Metric):
20 mm
25 mm
Surface Treatment:
Non-treated
Nunclon Delta
Unit Size:
Pack of 16
Pack of 20

Product Code. 10059030

Brand: Thermo Scientific™ 166508

Only null left
Add to basket
Add to basket

Screen large numbers of colonies for cloning experiments, culturing bacteria, fungi and agar diffusion assays using Thermo Scientific™ Nunc™ Squre BioAssay dishes.

Available in cell culture-treated and non-treated surfaces, the versatile Thermo Scientific™ Nunc™ Square BioAssay dishes are useful for screening large numbers of colonies, culturing bacteria and fungi, or providing a contained environment for other culture vessels.

  • Use as a moisture chamber for incubation on Nunc MicroWell™ Plates
  • Lined with filter paper it is ideal as an incubation chamber for in situ hybridization
  • Optically clear and compatible with automated robotic instrumentation

Untreated Dishes

  • Ideal for screening libraries or isolating clones
  • Useful for screening large numbers of colonies in cloning experiments
  • Can be used for culturing of bacteria and fungi as well as a moisture chamber for incubation of Nunc MicroWell Plates
  • Suitable in agar diffusion assay
  • Lined with wet filter paper, it is also useful as an incubation chamber for in situ hybridization
  • Large growth area
  • Designed for genomic screen and colony picking
TRUSTED_SUSTAINABILITY

Specifications

Bottom Shape Flat
Capacity (Metric) 135 mL
Color Clear
Culture Area 500 sq. cm
Shape Square
Sterility Sterile
Surface Treatment Nunclon Delta
Type Multi-Well Plate
Volume (Metric) Working 135 mL
No. per Case 16
No. per Pack 4
Height (Metric) 25 mm
Height (English) 25 in.
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.