missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ Tris-Acetate Mini Protein Gels, 7%, 1.0–1.5 mm

Invitrogen NuPAGE Tris-Acetate protein gels provide excellent separation of large molecular weight proteins. They are high performance polyacrylamide gels that simulate the denaturing or native conditions of the traditional laemmli system.

£155.00 - £165.00

Specifications

For Use With (Equipment) Mini Gel Tank, XCell SureLock Mini-Cell
Gel Percentage 7%
Gel Size Mini
Gel Type Tris-Acetate
Recommended Applications Denaturing, Native
View More Specs

Products 5
Product Code Brand Wells Gel Thickness Price Quantity & Availability  
Product Code Brand Wells Gel Thickness Price Quantity & Availability  
12035655
View Documents Promotion Details
Invitrogen™
EA0355BOX
10-well 1.0 mm
£155.00
Pack of 10
Estimated Shipment: 12-12-2024
to see stock.
Add to basket
 
12045655
View Documents Promotion Details
Invitrogen™
EA03552BOX
12-well 1.0 mm
£155.00
Pack of 10
Estimated Shipment: 06-12-2024
to see stock.
Add to basket
 
12055655
View Documents Promotion Details
Invitrogen™
EA03555BOX
15-well 1.0 mm
£155.00
Pack of 10
Estimated Shipment: 04-12-2024
to see stock.
Add to basket
 
12065655
View Documents Promotion Details
Invitrogen™
EA0358BOX
10-well 1.5 mm
£165.00
Pack of 10
Estimated Shipment: 12-12-2024
to see stock.
Add to basket
 
12075655
View Documents Promotion Details
Invitrogen™
EA03585BOX
15-well 1.5 mm
£165.00
Pack of 10
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
Description

Description

Invitrogen NuPAGE Tris-Acetate protein gels provide excellent separation of large molecular weight proteins. They are high performance polyacrylamide gels that simulate the denaturing or native conditions of the traditional laemmli system. The unique buffer formulation maintains a low operating pH during electrophoresis, resulting in superior resolution of high molecular weight proteins compared to traditional Tris-glycine SDS-PAGE gels.

Features of NuPAGE Tris-Acetate gels:
High resolution—gels offer optimal separation of high molecular weight proteins
Better protein integrity—sample preparation process has been optimized to help preserve proteins
Longer self life—gels can be stored for at least eight months

Choose the right NuPAGE Tris-Acetate gel for your protein separation
Obtain optimal separation of your high molecular weight proteins by choosing the right combination of gel and running buffer. NuPAGE Tris-Acetate protein gels come in a polyacrylamide concentration of 7% and a 3–8% gradient. Gels come in two sizes: mini (8 cm x 8 cm) or midi (8.7 cm x 13.3 cm) and either 1.0 mm (mini and midi gels) or 1.5 mm (mini gel format only) in thickness. NuPAGE Tris-Acetate gels also come in multiple well formats. Mini gels can be run using our XCell SureLock Mini-Cell (EI0001) or Mini Gel Tank (A25977). Midi gels can be run using our XCell4 SureLock Midi-Cell (WR0100) or conveniently with the Bio-Rad Criterion™ Cell using our adapters (WA0999).

Run your proteins in native or denatured form
NuPAGE Tris-Acetate protein gels do not contain SDS and can be used to separate proteins in native or denatured form. For denatured proteins, we recommend using NuPAGE LDS Sample Buffer (NP0007) and NuPAGE Tris-Acetate SDS Running Buffer (LA0041). For native proteins, we recommend using Novex Tris-Glycine Native Sample Buffer (LC2673) and Novex Tris-Glycine Native Running Buffer (LC2672).

For transfer of proteins to a membrane, we recommend using NuPAGE Transfer Buffer (NP00061) for traditional wet transfer using the XCell II Blot Module (EI9051) or the Mini Blot Module (B1000). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).

Specifications

Specifications

Mini Gel Tank, XCell SureLock Mini-Cell
Mini
Denaturing, Native
Denaturing, Native
NuPAGE™
Wet Ice
8 cm
7%
Tris-Acetate
36 to 500 kDa
Molecular Weight
10 Gels/Box
Store at 2°C to 8°C. Do not freeze.
8 cm
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 8 Months

SDS
Documents

Documents

Product Certifications