Learn More
Invitrogen™ NuPAGE™ Midi Tris-Acetate Resupply Pack, 3 to 8%
The NuPAGE Tris-Acetate Resupply Pack simplifies your large molecular weight protein separation workflow. This pack, sufficient for 100 gel runs, includes precast midi gels, running buffer, and protein ladders.
£1773.00
Specifications
Content And Storage | • NuPAGE Tris-Acetate gels, 10 x 10 gels • NuPAGE Tris-Acetate Running Buffer (20X), 5 bottles • HiMark Pre-stained Protein Standard (30 to 460 kDa), 4 vials |
---|---|
For Use With (Equipment) | SureLock Tandem Midi Gel Tank, XCell4 SureLock Midi-Cell |
Gel Percentage | 3 to 8% |
Gel Size | Midi |
Gel Type | Tris-Acetate |
Product Code | Brand | Wells | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Wells | Price | Quantity & Availability | |||||
30054930
|
Invitrogen™
STM4006-RS10 |
20-well |
£1773.00
Each |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30056178
|
Invitrogen™
STM4007-RS10 |
26-well |
£1773.00
Each |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The NuPAGE Tris-Acetate Resupply Pack simplifies your large molecular weight protein separation workflow. This pack, sufficient for 100 gel runs, includes precast midi gels, running buffer, and protein ladders. NuPAGE Tris-Acetate gels offer high-resolution protein separation while maintaining protein integrity. Designed for optimal separation and transfer they are ideal for researchers investigating large molecular weight proteins. With a long shelf life this pack is a reliable and convenient solution for your laboratory needs.
The NuPAGE Tris-Acetate Resupply Pack contains:
- NuPAGE Tris-Acetate gels (10 boxes, 100 gels)
- NuPAGE Tris-Acetate Running Buffer for 100 gel runs, 20X (5 bottles, Cat. No. LA0041)
- HiMark Pre-stained Protein Standard for 100 gel runs, 30 to 460 kDa (4 vials, Cat. No. LC5699)
About NuPAGE Tris-Acetate Gels
Invitrogen NuPAGE Tris-Acetate protein gels are precast polyacrylamide gels specifically engineered to provide excellent separation of large molecular weight proteins. These gels do not contain SDS, enabling you to run your proteins in native or denatured form, offering maximum flexibility for diverse protein electrophoresis needs. The unique buffer formulation of these gels maintains a low operating pH during electrophoresis, resulting in superior resolution of proteins when compared to traditional Tris-glycine SDS-PAGE gels.
Not only do NuPAGE Tris-Acetate gels offer high-resolution separation, but they are also designed to ensure optimal transfer of larger molecular weight proteins, making them a preferred choice for researchers performing western blotting. These gels represent a significant advancement over traditional gels, providing both superior performance and flexibility for protein gel electrophoresis and transfer. Additionally, midi format gels offer increased sample throughput per gel, enhancing efficiency, increasing reproducibility, and decreasing cost per sample.
Separation
For running Invitrogen protein gels, you have the flexibility to use either the SureLock Tandem Midi Gel Tank (STM1001) or the XCell4 SureLock Midi-Cell (WR0100). Both systems are designed to provide optimal performance. The SureLock Tandem offers rapid and efficient electrophoresis of midi gels in a leak-free system, with minimal buffer usage. The two independent chambers allow for electrophoresis of one or two gels simultaneously, conserving buffer and reducing waste. Alternatively, the XCell4 SureLock Midi-Cell is a robust and efficient system for midi-gel electrophoresis. Its standout feature is its capacity to simultaneously run up to four midi gels, enabling high-throughput protein analysis with enhanced efficiency and convenience. Additionally, the SureLock Tandem Midi Gel Tank is conveniently available through Invitrogen Precast Gel Welcome Packs along with both sample buffer and reducing agent, providing an all-in-one solution for your electrophoresis needs.
Transfer
For transferring your proteins to a membrane, we recommend using for NuPAGE Transfer Buffer (NP0006) for traditional wet transfer using the SureLock Tandem Midi Blot Module (STM2001). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001).
Specifications
• NuPAGE Tris-Acetate gels, 10 x 10 gels • NuPAGE Tris-Acetate Running Buffer (20X), 5 bottles • HiMark Pre-stained Protein Standard (30 to 460 kDa), 4 vials |
|
3 to 8% | |
Tris-Acetate | |
36 to 500 kDa | |
1 Kit |
SureLock Tandem Midi Gel Tank, XCell4 SureLock Midi-Cell | |
Midi | |
Protein | |
Denaturing, Native |
For Research Use Only. Not for use in diagnostic procedures.