missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ Tris-Acetate Resupply Pack, 3 to 8%, WedgeWell™ format

The NuPAGE Tris-Acetate Resupply Pack, Wedgewell format simplifies your large molecular weight protein separation workflow. This pack, sufficient for 100 gel runs, includes precast mini gels, running buffer, and protein ladders. NuPAGE Tris-Acetate gels offer high-resolution protein separation while maintaining protein integrity.

£1674.00

Specifications

Content And Storage • NuPAGE Tris-Acetate gels, 10 x 10 gels
• NuPAGE Tris-Acetate Running Buffer (20X), 4 bottles
• HiMark Pre-stained Protein Standard (30 to 460 kDa), 4 vials
For Use With (Equipment) Mini Gel Tank, XCell SureLock Mini-Cell
Gel Percentage 3 to 8%
Gel Size Mini
Gel Type Tris-Acetate
View More Specs

Products 3
Product Code Brand Wells Price Quantity & Availability  
Product Code Brand Wells Price Quantity & Availability  
30132194
View Documents Promotion Details
Invitrogen™
TA03810A-RS10
10-well
£1674.00
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30131431
View Documents Promotion Details
Invitrogen™
TA03810B-RS10
12-well
£1674.00
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30131716
View Documents Promotion Details
Invitrogen™
TA03810C-RS10
15-well
£1674.00
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The NuPAGE Tris-Acetate Resupply Pack, Wedgewell format simplifies your large molecular weight protein separation workflow. This pack, sufficient for 100 gel runs, includes precast mini gels, running buffer, and protein ladders. NuPAGE Tris-Acetate gels offer high-resolution protein separation while maintaining protein integrity. Designed for optimal separation and transfer they are ideal for researchers investigating large molecular weight proteins. With a long shelf life this pack is a reliable and convenient solution for your laboratory needs.

The NuPAGE Tris-Acetate Wedgewell Resupply Pack simplifies your large molecular weight protein separation workflow. This pack, sufficient for 100 gel runs, includes precast mini gels, running buffer, and protein ladders. NuPAGE Tris-Acetate gels offer high-resolution protein separation while maintaining protein integrity. Designed for optimal separation and transfer they are ideal for researchers investigating large molecular weight proteins. With a long shelf life this pack is a reliable and convenient solution for your laboratory needs.

Resupply Pack contents

  • NuPAGE Tris-Acetate WedgeWell gels (10 boxes, 100 gels)
  • NuPAGE Tris-Acetate Running Buffer for 100 gel runs, 20X (4 bottles, Cat. No. LA0041)
  • HiMark Pre-stained Protein Standard for 100 gel runs, 30 to 460 kDa (4 vials, Cat. No. LC5699)

About NuPAGE Tris-Acetate Gels

Invitrogen NuPAGE Tris-Acetate protein gels are precast polyacrylamide gels specifically engineered to provide excellent separation of large molecular weight proteins. These gels do not contain SDS, enabling you to run your proteins in native or denatured form, offering maximum flexibility for diverse protein electrophoresis needs. The unique buffer formulation of these gels maintains a low operating pH during electrophoresis, resulting in superior resolution of proteins when compared to traditional Tris-glycine SDS-PAGE gels.

Not only do NuPAGE Tris-Acetate gels offer high-resolution separation, but they are also designed to ensure optimal transfer of larger molecular weight proteins, making them a preferred choice for researchers performing western blotting. These gels represent a significant advancement over traditional gels, providing both superior performance and flexibility for protein gel electrophoresis and transfer.

Separation

For running Invitrogen protein gels, you have the flexibility to use either the Mini Gel Tank (A25977) or the XCell SureLock Mini-Cell (EI0001). Both systems are designed to provide optimal performance.. Additionally, the Mini Gel Tank is conveniently available through Invitrogen Precast Gel Welcome Packs along with both sample buffer and reducing agent, providing an all-in-one solution for your electrophoresis needs.

Transfer

For transferring your proteins to a membrane, we recommend using for NuPAGE Transfer Buffer (NP0006) for traditional wet transfer using the Mini Blot Module (B1000). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001).

Convenient to use

NuPAGE Tris-Acetate WedgeWell-format gels have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0-mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier.

Specifications

Specifications

• NuPAGE Tris-Acetate gels, 10 x 10 gels
• NuPAGE Tris-Acetate Running Buffer (20X), 4 bottles
• HiMark Pre-stained Protein Standard (30 to 460 kDa), 4 vials
3 to 8%
Tris-Acetate
40 to 500 kDa
1 Kit
Mini Gel Tank, XCell SureLock Mini-Cell
Mini
Protein
Denaturing, Native
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.