Learn More
Thermo Scientific™ Nutrient Agar (Slope)
Cultivate non-fastidious microorganisms for testing food, milk and dairy products using these nutritious, ready prepared slopes.
Brand: Thermo Scientific™ BO0336E
40.18 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Code : 65
Additional Details : Weight : 0.50000kg
Description
Cultivate non-fastidious microorganisms for testing food, milk and dairy products with Thermo Scientific™ Oxoid™ Nutrient Broth Slopes. This non-selective, fluid medium contains beef extracts combined with peptone, yeast extracts and sodium chloride to provide the nitrogen compounds, carbon, vitamins and some trace ingredients necessary for the growth of bacteria.
Remove the need for autoclaving and dispensing with convenient, pre-filled and ready prepared Slopes of Nutrient Broth.
- Suitable for general cultivation of less fasidious microorganisms.
- Convenient: the pre-filled slopes are ready for immediate use.
- Simple: easy-to-use with minimum training.
- Long shelf life: up to 12 months.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Straw, clear | |
Nutrient Agar (Slope) | |
United Kingdom | |
10.0g + 0.3g | |
24/Pk. |
2 to 25°C | |
Agar | |
Agar | |
Twenty four universal bottles with screw cap closures in a box. |