missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Invitrogen Collibri Stranded RNA Library Prep Kit

The Invitrogen Collibri Stranded RNA Library Preparation Kit Modules are designed for a custom build of next-generation sequencing (NGS) workflow on Illumina systems. The modules originally support whole-transcriptome sequencing (human, mouse, rat) and mRNA sequencing (compatible with all species) applications.

Brand:  Thermo Scientific™ A39117096

 View more versions of this product

Product Code. 17808115

  • £3145.00 / Pack of 96

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Invitrogen Collibri Stranded RNA Library Prep Kit for Illumina Systems is designed for the robust construction of cDNA libraries for strand-specific RNA sequencing on Illumina next-generation sequencing (NGS) systems. The modules bring together the superior features of SuperScript IV Reverse Transcriptase, Dynabeads magnetic particles, and Platinum SuperFi DNA Polymerase, enabling users to obtain high-quality sequencing-ready libraries.

The Invitrogen Collibri Stranded RNA Library Prep Kit for Illumina Systems is designed for the robust construction of cDNA libraries for strand-specific RNA sequencing on Illumina next-generation sequencing (NGS) systems. The modules bring together the superior features of SuperScript IV Reverse Transcriptase, Dynabeads magnetic particles, and Platinum SuperFi DNA Polymerase, enabling users to obtain high-quality, sequencing-ready libraries.

This Collibri Stranded RNA Library Prep Kit Modules supports fast library preparation within 4.5 hours for mRNA or whole transcriptome libraries with 1–25 ng of mRNA-enriched or ribodepleted RNA sample input. Multiplexing of libraries can be carried out using up to 96 single-indexed or unique dual-indexed primers, enabling single-read or paired-end sequencing. The kit is suitable for various-quality RNA samples, including FFPE samples.

Visual cues of process integrity
For maximum convenience, visual cues of process integrity are included throughout the library preparation workflow. Monitor progress of library generation using visual feedback from dyes in critical reagents. The reaction mix changes color in response to the addition of critical components at each step to ensure that library preparation has the highest chance of success. The inert dyes do not interfere with enzymatic reactions and do not compromise sequencing results.

How it works
Full-length Illumina-compatible adaptors are introduced by PCR amplification using Platinum SuperFi DNA Polymerase to construct single-indexed or unique dual-indexed libraries compatible with single-read or paired-end sequencing. The kit contains 24 or 96 either single-indexed barcoded PCR primers (i7) pre-mixed with a universal i5 primer or unique dual-indexed PCR primers that enable multiplexing of up to 96 libraries. Optimized cleanup steps efficiently remove residual primers and adapter/primer dimers while preserving high library yields.

A related product, the Collibri Library Quantification Kit, is recommended for qPCR-based quantifications of libraries before proceeding to sequencing.

Applications
The Collibri Stranded RNA Library Prep Kit is recommended for:
• Gene expression studies
• Alternative splicing analysis
• Non-coding RNA detection and discovery
• Identification of alternative polyadenylation sites
• Gene fusion detection
• Detection of transcript isoforms

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at -20°C
Kit
Collibri™
96 preps
Fragmentation, Indexing, Ligation, PCR Amplification, Reverse Transcription
RNA sequencing; whole transcriptome; mRNA sequencing; small RNA sequencing
Stranded RNA Library Prep Kit
96 preps
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.