missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NVP 231, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent, selective and reversible ceramide kinase (CerK) inhibitor (IC50 = 12 nM in vitro). Displays selectivity for CerK over sphingosine kinases (IC50 ≥100 μM) and other lipid kinases.
Specifications
Quantity | 10 mg |
Formula Weight | 431.55 |
Percent Purity | >98% |
Chemical Name or Material | NVP 231 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'