missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NVP 231
GREENER_CHOICE

Product Code. 18015442
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18015442

Brand: Tocris Bioscience™ 3960/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent, selective and reversible CerK inhibitor

Potent, selective and reversible ceramide kinase (CerK) inhibitor (IC50 = 12 nM in vitro). Displays selectivity for CerK over sphingosine kinases (IC50 ≥100 μM) and other lipid kinases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 362003-83-6
Molecular Formula C25H25N3O2S

Specifications

Quantity 50 mg
Formula Weight 431.55
Purity >98%
Molecular Weight (g/mol) 431.554
Chemical Name or Material NVP 231
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.