Learn More
Thermo Scientific™ NY/T Method QuEChERS Clean-up Kit for 15 g Samples, General Fruits and Vegetables
Use Thermo Scientific™ QuEChERS NY/T 1380-2007 Method Clean-up Kits for accurate determination of 51 specific pesticide residues in fruits and vegetables.
Brand:  Thermo Scientific™ S2-15-GFV-NYT-KIT
Product Code. 17349842
  • £120.00 / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Use Thermo Scientific™ QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) NY/T 1380-2007 Method Clean-up Kits for accurate and efficient determination of 51 specific pesticide residues in fruits and vegetables. Our QuEChERS NY/T 1380-2007 Method Clean-up Kits are ideal for preparing samples prior to GC-MS and can be automated for higher throughput.
The NY/T 1380-2007 Method supports voluntary testing of professional and industrial standards for agriculture in China. Thermo Scientific™ QuEChERS NY/T 1380-2007 Method Clean-up Kits make sample preparation straight-forward and efficient, allowing for determination of pesticide residues in fruit and vegetable matrices.
We offer a clean-up/dispersive SPE kit composed of pre-filled 15 mL tubes for use with general fruit and vegetable matrices.
What is the purpose of the chemicals used in the NY/T 1380-2007 Method?
  • Acetonitrile: used to perform single-phase extraction on a sample
  • Anhydrous MgSO4: induces phase separation between water content in sample and acetonitrile layer during liquid-liquid partitioning step; removes remaining water from sample during dispersive solid phase extraction (dSPE) step
  • PSA: primary secondary amine, used to remove polar matrix components, citric co-extractives, and free fatty acids, from sample during dispersive solid phase extraction (dSPE) step
  • C18: removes fats, sterols, and other non-polar interferences from sample
TRUSTED_SUSTAINABILITY
Specifications
Specifications
QuEChERS NY/T 1380-2007 Method Clean-up Kit; 100 mg PSA, 100 mg C18, 300 mg Anhydrous MgSO4 prefilled in 15 mL Tubes; 50/Pk.
Salts in Prefilled Tubes
300 mg Anhydrous MgSO4, 100 mg PSA, 100 mg C18
General Fruits and Vegetables
QuEChERS Clean-up Kit
QuEChERS sample preparation for NY/T 1380-2007 Method
15 g sample
15 mL