missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Nylon Biopsy Bags
Ensure unrestricted fluid movement around tissue with Epredia™ Nylon Biopsy Bags.
£170.00 - £728.00
Product Code | Brand | Size | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Quantity | Price | Quantity & Availability | ||||
12066789
|
Epredia™
6774009 |
Medium | 1000/Pk. |
£537.00
Pack of 1000 |
Estimated Shipment: 09-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12076789
|
Epredia™
6774010 |
Small | 1000/Pk. |
£728.00
Pack of 1000 |
Estimated Shipment: 11-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12637946
|
Epredia™
6774011 |
Large | 500/Pk. |
£349.00
Pack of 500 |
Estimated Shipment: 11-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12657946
|
Epredia™
6774012 |
Medium | 200/Pk. |
£170.00
Pack of 200 |
Estimated Shipment: 11-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11932355
|
Epredia™
6774014 |
Small | 200/Pk. |
£212.00
Pack of 200 |
Estimated Shipment: 11-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12066699
|
Epredia™
6774015 |
Small | 500/Pk. |
£361.20
Pack of 500 |
Estimated Shipment: 11-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12056699
|
Epredia™
6774013 |
Medium | 500/Pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Reduce risk of small specimen loss
- Reduce reagent carryover and pressure artifact often found with sponges
- Remove specimen rapidly with convenient peel-open bag
Dimensions:
- Small Biopsy Bags measure 1.2 × 1.7 in. (31 × 43mm)
- Medium Biopsy Bags measure 1.8 × 2.6 in. (45 × 65mm)
- Large Biopsy Bags measure 3 × 3.5 in. (75 × 85mm)
Specifications
Biopsy Bag | |
Nylon | |
Blue |