Learn More
Azpack™ Nylon Cable Tie Bases
Adhere to flat surfaces with self-stick backing. Azpack™ Nylon Cable Tie Bases provide a surface to tie down cables and other objects.
Brand: Azpack™ 11378093
18.64 GBP valid until 2024-07-31
Use promo code "21640" to get your promotional price.
Code : 03
Additional Details : Weight : 0.06480kg
Description
- Accepts cable zip ties between 2.5 and 3.6mmW.
- Self-adhesive
- Rigid nylon construction
Specifications
Natural | |
UL94 Class V2 | |
20 x 20 mm | |
20 mm |
100 Pk. | |
Nylon | |
20 mm |