missing translation for 'onlineSavingsMsg'
Learn More
Ansell Edmont™ Marigold™ Picolon Confort™ Polyamide Gloves Product Code.: 12382209
Add to basket

Ansell Edmont™ Marigold™ Picolon Confort™ Polyamide Gloves

Product Code. 12382209
Medium
Click to view available options
Size:
Medium
Large
X-Large

Product Code. 12382209

Brand: Ansell Edmont™ C357080

Edge
Add to basket
Add to basket

Providing good resistance to abrasion and small blade cut hazards. Ansell Edmont™ Marigold™ Picolon Confort™ Polyamide Gloves are ideal for the handling and assembly of slightly sharp and slippery objects.

  • Dots on palm and fingers for better grip
  • Textile liner made of high elasticity polyamide thread
  • Fine, stretchable and comfortable stitches
  • Knitted with a seamless non-stop process
  • Lint-free
TRUSTED_SUSTAINABILITY

Specifications

Construction Seamless Knit
Coating Material PVC
Color White with Blue Dots
Size Medium
Glove Cuff Style Knit
Liner Material Polyamide
Resistant To Abrasion and small cuts
Length (Metric) 260 mm
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131

    Expires2025-06-08

    New Offer
    Shop Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell Edmont™ Marigold™ Picolon Confort™ Polyamide Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.