missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Nylon Membrane Filter

Naturally hydrophilic membrane designed to wet out evenly and retain its superior strength during use in general filtration or medical assays.

Brand:  Fisherbrand™ 0462A00021

 View more versions of this product

Product Code. 11714516

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Naturally hydrophilic membrane
  • Designed to wet out evenly and retain its superior strength during use in general filtration or medical assays
  • Flexible, durable and tear resistant
  • Autoclaved at 135°C
  • Effective chemical compatibility with aqueous and alcoholic solutions and solvents
Specifications

Specifications

Autoclavable
White
Nylon
Non-sterile
Sterilizing filtration of aqueous solutions, organic solvents, Bacterial and particles removal (blood Glucose, Serum Cholesterol, drugs)
Hydrophilic
Plain Disc
13 mm
3.4bar
Microfilter
0.22 μm
9.9mL/min./cm2
1.7L/min./cm2
100 Pack
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Nylon Membrane Filter > Pore Size: 0.22 μm, Diameter: 13 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.