missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Nylon Membrane Filter

Naturally hydrophilic membrane designed to wet out evenly and retain its superior strength during use in general filtration or medical assays.

Brand:  Fisherbrand™ 0464A00023

62.17 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.


 View more versions of this product

Product Code. 11765315

  • £79.20 / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Naturally hydrophilic membrane
  • Designed to wet out evenly and retain its superior strength during use in general filtration or medical assays
  • Flexible, durable and tear resistant
  • Autoclaved at 135°C
  • Effective chemical compatibility with aqueous and alcoholic solutions and solvents
Specifications

Specifications

Autoclavable
White
Nylon
Non-sterile
Sterilizing filtration of aqueous solutions, organic solvents, Bacterial and particles removal (blood Glucose, Serum Cholesterol, drugs)
Hydrophilic
Plain Disc
1.85 in.
2bar
Microfilter
0.45 μm
26.9mL/min./cm2
3.2L/min./cm2
100 Pack
Yes
47 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

    Days

    73

    Hours

    7

    Minutes

    10

    Seconds

    22
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Nylon Membrane Filter > Pore Size: 0.45 μm, Diameter: 47 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.