missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Nylon Membrane Filter
Naturally hydrophilic membrane designed to wet out evenly and retain its superior strength during use in general filtration or medical assays.
£47.01 - £84.60
Specifications
Autoclavable | Autoclavable |
---|---|
Color | White |
Filter Type | Microfilter |
Material | Nylon |
Sterility | Non-sterile |
Product Code | Brand | Pore Size | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Diameter (Metric) | Price | Quantity & Availability | |||||
11714516
|
Fisherbrand™
0462A00021 |
0.22 μm | 13 mm | N/A | ||||||
11735315
|
Fisherbrand™
0462A00022 |
0.22 μm | 25 mm |
£62.55
Pack of 100 |
Estimated Shipment: 16-04-2025
Log in to see stock. |
|||||
11724516
|
Fisherbrand™
0464A00021 |
0.45 μm | 13 mm | N/A | ||||||
11745315
|
Fisherbrand™
0462A00023 |
0.22 μm | 47 mm |
£84.60
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||||
11755315
|
Fisherbrand™
0464A00022 |
0.45 μm | 25 mm |
£74.57
Pack of 100 |
Estimated Shipment: 16-04-2025
Log in to see stock. |
|||||
11765315
|
Fisherbrand™
0464A00023 |
0.45 μm | 47 mm |
£81.97
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||||
Description
- Naturally hydrophilic membrane
- Designed to wet out evenly and retain its superior strength during use in general filtration or medical assays
- Flexible, durable and tear resistant
- Autoclaved at 135°C
- Effective chemical compatibility with aqueous and alcoholic solutions and solvents
Specifications
Autoclavable | |
Microfilter | |
Non-sterile | |
Hydrophilic | |
Plain Disc |
White | |
Nylon | |
Sterilizing filtration of aqueous solutions, organic solvents, Bacterial and particles removal (blood Glucose, Serum Cholesterol, drugs) | |
100 Pack | |
Yes |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Nylon Membrane Filter