missing translation for 'onlineSavingsMsg'
Learn More
Learn More
De Buyer™ Nylon Soft Spatulas
Scoop, spread and scrape ingredients using De Buyer™ Nylon Soft lSpatulas. The spatula can withstand temperatures up to 260°.
Brand: De Buyer™ 4740.24N
This item is not returnable.
View return policy
Description
- Thermoplasticized
- Up to 260 degrees maximum
- Plastic handle
Specifications
1 Ea. | |
Silicone | |
Plastic | |
240 mm |
Soft Spatula | |
260°C | |
Nylon |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
De Buyer™ Nylon Soft Spatulas > Length: 240mm; Silicone
Spot an opportunity for improvement?Share a Content Correction