missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Masterflex™ Nylon Threaded Fittings
£8.65 - £91.60
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
11770369
|
Masterflex™
30401-42 |
0.25 in. NPT(M) |
£27.60
Pack of 10 |
Estimated Shipment: 03-09-2024
Log in to see stock. |
|||||
11751084
|
Masterflex™
06349-61 |
0.25in. Tubing |
£41.75
Pack of 6 |
Estimated Shipment: 03-09-2024
Log in to see stock. |
|||||
11745768
|
Masterflex™
31701-91 |
0.125 in. NPT(M) |
£8.65
Pack of 10 |
Estimated Shipment: 03-09-2024
Log in to see stock. |
|||||
11780369
|
Masterflex™
30401-50 |
0.375 in. NPT(M) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11711104
|
Masterflex™
06349-52 |
0.375in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11791084
|
Masterflex™
06349-50 |
0.125in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11731104
|
Masterflex™
06349-54 |
0.75in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11701154
|
Masterflex™
06349-87 |
0.75 x 0.5in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Threaded Fitting | |
Nylon |