missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Nylon Ties

Product Code. 11736895
Click to view available options
Color:
Blue
Orange
Purple
Red
White
Yellow
Unit Size:
1 pack
This item is not returnable. View return policy

Product Code. 11736895

Brand: Brady™ 229961

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
  • Cross-laminated polyethylene
  • Matte finish

Specifications

Type Ties
Material Nylon
Color Purple
Width (Metric) 3.6 mm
Quantity 100/Pk.
For Use With (Application) Ideal for color-coding hard to mark items
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.