Learn More
Cole-Parmer™ Nylon Turbine Flowmeter
Brand:  Cole-Parmer™ 05610-01
Product Code. 11727338
  • £818.00 / Each
Explore more special offers
Specifications
Turbine Flowmeter
Nylon
1 in. NPT (F)
Low-cost solution to light-duty process applications
Two 3 V Lithium Batteries
110 mm
600 g
±5% (Reading)
150 psig
1 to 10 LPM
64 mm
±0.1%
60°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products