Learn More
O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 98%, Thermo Scientific Chemicals
£27.54 - £81.11
Chemical Identifiers
CAS | 94790-37-1 |
---|---|
Molecular Formula | C11H16F6N5OP |
Molecular Weight (g/mol) | 379.247 |
MDL Number | MFCD00075445 |
InChI Key | UQYZFNUUOSSNKT-UHFFFAOYSA-N |
Synonym | hbtu, o-benzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate, 2-1h-benzotriazole-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate, hbtu-reagent, o-benzotriazol-1-yl-tetramethyluronium hexafluorophosphate, hbtu o-benzotriazole-n,n,n',n'-tetramethyl-uronium-hexafluoro-phosphate, n,n,n',n'-tetramethyl-o-1h-benzotriazol-1-yl uronium hexafluorophosphate, o-1h-benzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate, o-benzotriazole-n,n,n',n'-tetramethyluronium hexafluorophosphateShow More |
PubChem CID | 2733084 |
IUPAC Name | [benzotriazol-1-yloxy(dimethylamino)methylidene]-dimethylazanium;hexafluorophosphate |
SMILES | CN(C)C(=[N+](C)C)ON1C2=CC=CC=C2N=N1.F[P-](F)(F)(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11435627
|
Thermo Scientific Alfa Aesar
B23597.06 |
5 g |
£27.54
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11445627
|
Thermo Scientific Alfa Aesar
B23597.14 |
25 g |
£44.22
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11455627
|
Thermo Scientific Alfa Aesar
B23597.22 |
100 g |
£81.11
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate is used as a coupling reagent for solid-phase peptide synthesis. It has been shown to effectively suppress racemization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsO-(1H-Benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium hexafluorophosphate is used as a coupling reagent for solid-phase peptide synthesis. It has been shown to effectively suppress racemization.
Solubility
Soluble in acetonitrile.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Chemical Identifiers
94790-37-1 | |
379.247 | |
UQYZFNUUOSSNKT-UHFFFAOYSA-N | |
2733084 | |
CN(C)C(=[N+](C)C)ON1C2=CC=CC=C2N=N1.F[P-](F)(F)(F)(F)F |
C11H16F6N5OP | |
MFCD00075445 | |
hbtu, o-benzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate, 2-1h-benzotriazole-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate, hbtu-reagent, o-benzotriazol-1-yl-tetramethyluronium hexafluorophosphate, hbtu o-benzotriazole-n,n,n',n'-tetramethyl-uronium-hexafluoro-phosphate, n,n,n',n'-tetramethyl-o-1h-benzotriazol-1-yl uronium hexafluorophosphate, o-1h-benzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate, o-benzotriazole-n,n,n',n'-tetramethyluronium hexafluorophosphateShow More | |
[benzotriazol-1-yloxy(dimethylamino)methylidene]-dimethylazanium;hexafluorophosphateShow More |
Specifications
94790-37-1 | |
C11H16F6N5OP | |
7328329 | |
Soluble in acetonitrile. | |
CN(C)C(=[N+](C)C)ON1C2=CC=CC=C2N=N1.F[P-](F)(F)(F)(F)F | |
379.247 | |
379.25 | |
O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate |
∼200°C (decomposition) | |
MFCD00075445 | |
hbtu, o-benzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate, 2-1h-benzotriazole-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate, hbtu-reagent, o-benzotriazol-1-yl-tetramethyluronium hexafluorophosphate, hbtu o-benzotriazole-n,n,n',n'-tetramethyl-uronium-hexafluoro-phosphate, n,n,n',n'-tetramethyl-o-1h-benzotriazol-1-yl uronium hexafluorophosphate, o-1h-benzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate, o-benzotriazole-n,n,n',n'-tetramethyluronium hexafluorophosphate | |
UQYZFNUUOSSNKT-UHFFFAOYSA-N | |
[benzotriazol-1-yloxy(dimethylamino)methylidene]-dimethylazanium;hexafluorophosphate | |
2733084 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 423-020-5
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.