Learn More
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%, C7H4F5NO.HCl, CAS Number-57981-02-9
£25.40 - £275.00
Chemical Identifiers
CAS | 57981-02-9 |
---|---|
Molecular Formula | C7H5ClF5NO |
Molecular Weight (g/mol) | 249.57 |
MDL Number | MFCD00012953 |
InChI Key | HVMVKNXIMUCYJA-UHFFFAOYSA-N |
Synonym | o-2,3,4,5,6-pentafluorobenzyl hydroxylamine hydrochloride, florox reagent, o-perfluorophenyl methyl hydroxylamine hydrochloride, hydroxylamine, o-pentafluorophenyl methyl-, hydrochloride, o-pentafluorobenzyl hydroxylamine hydrochloride, o-2,3,4,5,6-pentafluorophenyl methyl hydroxylamine hydrochloride, oxy 2,3,4,5,6-pentafluorophenyl methyl amine, chloride, pfbha.hcl, acmc-1b0zv |
PubChem CID | 122307 |
IUPAC Name | O-[(2,3,4,5,6-pentafluorophenyl)methyl]hydroxylamine;hydrochloride |
SMILES | [H+].[Cl-].NOCC1=C(F)C(F)=C(F)C(F)=C1F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15626804
|
Thermo Scientific Acros
186712500 |
250 mg |
£25.40
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15606804
|
Thermo Scientific Acros
186710010 |
1 g |
£71.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15616804
|
Thermo Scientific Acros
186710050 |
5 g |
£275.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
57981-02-9 | |
249.57 | |
HVMVKNXIMUCYJA-UHFFFAOYSA-N | |
122307 | |
[H+].[Cl-].NOCC1=C(F)C(F)=C(F)C(F)=C1F |
C7H5ClF5NO | |
MFCD00012953 | |
o-2,3,4,5,6-pentafluorobenzyl hydroxylamine hydrochloride, florox reagent, o-perfluorophenyl methyl hydroxylamine hydrochloride, hydroxylamine, o-pentafluorophenyl methyl-, hydrochloride, o-pentafluorobenzyl hydroxylamine hydrochloride, o-2,3,4,5,6-pentafluorophenyl methyl hydroxylamine hydrochloride, oxy 2,3,4,5,6-pentafluorophenyl methyl amine, chloride, pfbha.hcl, acmc-1b0zv | |
O-[(2,3,4,5,6-pentafluorophenyl)methyl]hydroxylamine;hydrochloride |
Specifications
57981-02-9 | |
Authentic | |
C7H5ClF5NO | |
MFCD00012953 | |
Solubility in water: soluble. | |
[H+].[Cl-].NOCC1=C(F)C(F)=C(F)C(F)=C1F | |
249.57 | |
249.57 | |
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride |
227.0°C | |
97.5 to 102.5% (Argentometry) | |
C6F5CH2ONH2·HCl | |
o-2,3,4,5,6-pentafluorobenzyl hydroxylamine hydrochloride, florox reagent, o-perfluorophenyl methyl hydroxylamine hydrochloride, hydroxylamine, o-pentafluorophenyl methyl-, hydrochloride, o-pentafluorobenzyl hydroxylamine hydrochloride, o-2,3,4,5,6-pentafluorophenyl methyl hydroxylamine hydrochloride, oxy 2,3,4,5,6-pentafluorophenyl methyl amine, chloride, pfbha.hcl, acmc-1b0zv | |
HVMVKNXIMUCYJA-UHFFFAOYSA-N | |
O-[(2,3,4,5,6-pentafluorophenyl)methyl]hydroxylamine;hydrochloride | |
122307 | |
98% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 261-057-7
RUO – Research Use Only