missing translation for 'onlineSavingsMsg'
Learn More
o-Benzoic sulfimide, 98+%, Thermo Scientific Chemicals Product Code.: 11476806

o-Benzoic sulfimide, 98+%, Thermo Scientific Chemicals

Product Code. 11476806
100 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11476806

Brand: Thermo Scientific Alfa Aesar B23938.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

o-Benzoic sulfimide is used as a pharmaceutic flavor aid. It is used in high performance liquid chromatographic method for the simultaneous separation and determination of acesulfame potassium, saccharine and aspartame.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Benzoic sulfimide is used as a pharmaceutic flavor aid. It is used in high performance liquid chromatographic method for the simultaneous separation and determination of acesulfame potassium, saccharine and aspartame.

Solubility
Soluble in water (3.3 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-07-2
Molecular Formula C7H5NO3S
Molecular Weight (g/mol) 183.181
MDL Number MFCD00005866
InChI Key CVHZOJJKTDOEJC-UHFFFAOYSA-N
Synonym saccharin, saccharine, o-sulfobenzimide, o-benzoic sulfimide, saccharimide, benzosulfimide, o-benzosulfimide, benzoic sulfimide, benzosulphimide, saccharinoseShow MoreShow Less
PubChem CID 5143
ChEBI CHEBI:32111
IUPAC Name 1,1-dioxo-1,2-benzothiazol-3-one
SMILES C1=CC=C2C(=C1)C(=O)NS2(=O)=O

Specifications

Melting Point 227°C to 230°C
Density 0.828
Quantity 100 g
Beilstein 6888
Merck Index 14,8311
Solubility Information Soluble in water (3.3g/L at 20°C).
Formula Weight 183.18
Percent Purity ≥98%
Chemical Name or Material o-Benzoic sulfimide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10241898 Certificate TypeCertificate of Analysis Date08/07/2022 Catalog NumberB23938.0E,B23938.22,B23938.36
Lot Number10151557 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23938.0E,B23938.22,B23938.36
Lot Number10224567 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23938.0E,B23938.22,B23938.36
Lot Number10115502 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23938.0E,B23938.22,B23938.36
Lot Number10205057 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23938.0E,B23938.22,B23938.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
o-Benzoic sulfimide, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.