missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-Benzyl-L-tyrosine, 98%, Thermo Scientific Chemicals
£43.30 - £374.00
Chemical Identifiers
CAS | 16652-64-5 |
---|---|
Molecular Formula | C16H17NO3 |
Molecular Weight (g/mol) | 271.32 |
MDL Number | MFCD00002605 |
InChI Key | KAFHLONDOVSENM-GGYSOQFKNA-N |
Synonym | o-benzyl-l-tyrosine, h-tyr bzl-oh, 4-benzyloxy-l-phenylalanine, s-2-amino-3-4-benzyloxy phenyl propanoic acid, o-benzyl-l-tyr, tyrosine, o-phenylmethyl, l-tyrosine benzyl ether, 2s-2-amino-3-4-benzyloxy phenyl propanoic acid, h-tyr bn-oh |
PubChem CID | 725701 |
IUPAC Name | (2S)-2-amino-3-(4-phenylmethoxyphenyl)propanoic acid |
SMILES | N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15440489
|
Thermo Scientific Alfa Aesar
H63675.06 |
5 g |
£43.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15450489
|
Thermo Scientific Alfa Aesar
H63675.14 |
25 g |
£136.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15460489
|
Thermo Scientific Alfa Aesar
H63675.22 |
100 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16652-64-5 | |
271.32 | |
KAFHLONDOVSENM-GGYSOQFKNA-N | |
725701 | |
N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(O)=O |
C16H17NO3 | |
MFCD00002605 | |
o-benzyl-l-tyrosine, h-tyr bzl-oh, 4-benzyloxy-l-phenylalanine, s-2-amino-3-4-benzyloxy phenyl propanoic acid, o-benzyl-l-tyr, tyrosine, o-phenylmethyl, l-tyrosine benzyl ether, 2s-2-amino-3-4-benzyloxy phenyl propanoic acid, h-tyr bn-oh | |
(2S)-2-amino-3-(4-phenylmethoxyphenyl)propanoic acid |
Specifications
259°C (decomposition) | |
White | |
MFCD00002605 | |
o-benzyl-l-tyrosine, h-tyr bzl-oh, 4-benzyloxy-l-phenylalanine, s-2-amino-3-4-benzyloxy phenyl propanoic acid, o-benzyl-l-tyr, tyrosine, o-phenylmethyl, l-tyrosine benzyl ether, 2s-2-amino-3-4-benzyloxy phenyl propanoic acid, h-tyr bn-oh | |
N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(O)=O | |
271.32 | |
271.31 | |
Powder |
16652-64-5 | |
C16H17NO3 | |
2659642 | |
KAFHLONDOVSENM-GGYSOQFKNA-N | |
(2S)-2-amino-3-(4-phenylmethoxyphenyl)propanoic acid | |
725701 | |
98% | |
O-Benzyl-L-tyrosine |
Safety and Handling
missing translation for 'einecsNumber' : 240-699-1
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only