missing translation for 'onlineSavingsMsg'
Learn More

o-Cresol, 99%, Thermo Scientific Chemicals

£31.56 - £377.88

Chemical Identifiers

CAS 95-48-7, 108-95-2
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00002226
InChI Key QWVGKYWNOKOFNN-UHFFFAOYSA-N
Synonym o-cresol, 2-hydroxytoluene, o-methylphenol, 2-cresol, phenol, 2-methyl, orthocresol, o-cresylic acid, o-oxytoluene, o-toluol, o-hydroxytoluene
PubChem CID 335
ChEBI CHEBI:28054
IUPAC Name 2-methylphenol
SMILES CC1=CC=CC=C1O
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10442865
View Documents Promotion Details
Thermo Scientific Acros
110551000
100 g Glass bottle
£31.56
100g
Please to purchase this item. Need a web account? Register with us today!
10346220
View Documents Promotion Details
Thermo Scientific
110555000
500 g Glass bottle
£37.08
500g
Please to purchase this item. Need a web account? Register with us today!
10498630
View Documents Promotion Details
Thermo Scientific Acros
110550010
1 kg Glass bottle
£56.18
1kg
Please to purchase this item. Need a web account? Register with us today!
10300054
View Documents Promotion Details
Thermo Scientific Acros
110550025
2.5 kg Glass bottle
£111.56
2.50kg
Please to purchase this item. Need a web account? Register with us today!
13488298
View Documents Promotion Details
Thermo Scientific Acros
110550100
10 kg Plastic drum
£377.88
10kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

95-48-7, 108-95-2
108.14
QWVGKYWNOKOFNN-UHFFFAOYSA-N
335
2-methylphenol
C7H8O
MFCD00002226
o-cresol, 2-hydroxytoluene, o-methylphenol, 2-cresol, phenol, 2-methyl, orthocresol, o-cresylic acid, o-oxytoluene, o-toluol, o-hydroxytoluene
CHEBI:28054
CC1=CC=CC=C1O

Specifications

95-48-7, 108-95-2
100.0
Amber to Colorless
191.0°C
Authentic
C7H8O
MFCD00002226
15,2564
o-cresol, 2-hydroxytoluene, o-methylphenol, 2-cresol, phenol, 2-methyl, orthocresol, o-cresylic acid, o-oxytoluene, o-toluol, o-hydroxytoluene
QWVGKYWNOKOFNN-UHFFFAOYSA-N
2-methylphenol
335
4.75 mPa.s (35°C)
99%
o-Cresol
98.5
30.0°C to 32.0°C
1.0400g/mL
81°C
98.5% min. (GC)
CH3C6H4OH
06,349
1.04
Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol,ether and chloroform
CC1=CC=CC=C1O
108.14
CHEBI:28054
108.14
Liquid or Low Melting Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 202-423-8

Recommended Storage : Darkens on storage and exposure to light and air

Product Identifier
  • o-Cresol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P302+P350-IF ON SKIN: Gently wash with plenty of soap and water.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only