missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals o-Cresolphthalein Complexone, pure

Product Code. 10269413
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2411-89-4
Molecular Formula C32H32N2O12
Molecular Weight (g/mol) 636.61
MDL Number MFCD00005911
InChI Key IYZPEGVSBUNMBE-UHFFFAOYSA-N
Synonym o-Cresolphthalexon, Phthalein Complexon
PubChem CID 75485
IUPAC Name 2-[({5-[1-(3-{[bis(carboxymethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-3-oxo-1,3-dihydro-2-benzofuran-1-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxymethyl)amino]acetic acid
SMILES CC1=CC(=CC(CN(CC(O)=O)CC(O)=O)=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC(O)=O)CC(O)=O)=C1

Specifications

Melting Point 181.0°C
Merck Index 15, 2566
Solubility Information Solubility in water: slightly soluble. Other solubilities: readily soluble in aqueous ammonia, readily soluble in organic solvents
Infrared Spectrum Authentic
Lambda Max 573 to 579nm (in 0.1N NaOH)
Formula Weight 636.6
Loss on Drying 10% max. (1 g, 105°C)
Grade Pure
Packaging Glass bottle
Suitability As indicator for complexometry: Passes Test
UV Spectrometry Complies
Color Brown to White
Quantity 100 g
Physical Form Powder
Chemical Name or Material o-Cresolphthalein Complexone, Pure
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'