missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals o-Cresolphthalein Complexone, pure

Product Code. p-2367921 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10285850

Brand: Thermo Scientific Chemicals 182440050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2411-89-4 | C32H32N2O12 | 636.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2411-89-4
Molecular Formula C32H32N2O12
Molecular Weight (g/mol) 636.61
MDL Number MFCD00005911
InChI Key IYZPEGVSBUNMBE-UHFFFAOYSA-N
Synonym o-Cresolphthalexon, Phthalein Complexon
PubChem CID 75485
IUPAC Name 2-[({5-[1-(3-{[bis(carboxymethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-3-oxo-1,3-dihydro-2-benzofuran-1-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxymethyl)amino]acetic acid
SMILES CC1=CC(=CC(CN(CC(O)=O)CC(O)=O)=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC(O)=O)CC(O)=O)=C1

Specifications

Melting Point 181.0°C
Merck Index 15, 2566
Solubility Information Solubility in water: slightly soluble. Other solubilities: readily soluble in aqueous ammonia, readily soluble in organic solvents
Infrared Spectrum Authentic
Lambda Max 573 to 579nm (in 0.1N NaOH)
Formula Weight 636.6
Loss on Drying 10% max. (1 g, 105°C)
Grade Pure
Packaging Glass bottle
Suitability As indicator for complexometry: Passes Test
UV Spectrometry Complies
Color Brown to White
Quantity 5 g
Physical Form Powder
Chemical Name or Material o-Cresolphthalein Complexone, Pure
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.