missing translation for 'onlineSavingsMsg'
Learn More

o-Cresolphthalein

Product Code. 11425353
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11425353

Brand: Thermo Scientific Alfa Aesar A12899.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 596-27-0 | C22H18O4 | 346.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 596-27-0
Molecular Formula C22H18O4
Molecular Weight (g/mol) 346.38
MDL Number MFCD00005912
InChI Key CPBJMKMKNCRKQB-UHFFFAOYSA-N
Synonym o-cresolphthalein, cresolphthalein, 3,3'-dimethylphenolphthalein, 1 3h-isobenzofuranone, 3,3-bis 4-hydroxy-3-methylphenyl, 3',3-dimethylphenolphthalein, o-cresolphalein, 3,3-bis 4-hydroxy-3-methylphenyl isobenzofuran-1 3h-one, unii-xou2d9049g, phenolphthalein, 3',3-dimethyl, 3,3-bis 4-hydroxy-m-tolyl phthalide
PubChem CID 68995
SMILES CC1=CC(=CC=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1

Specifications

Melting Point 223°C to 225°C
Quantity 50 g
Beilstein 310554
Merck Index 14,2576
IUPAC Name 3,3-bis(4-hydroxy-3-methylphenyl)-2-benzofuran-1-one
Formula Weight 346.39
Chemical Name or Material o-Cresolphthalein

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.