missing translation for 'onlineSavingsMsg'
Learn More

O-Cresolsulfonphthaleindi-(Methyl-Iminodiacetic Acid) Sodium Salt, Pure, special reagent for metals, Fisher Chemical™

Catalog Number 10040080
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
1611-35-4
C31H32N2O13S
672.66
11493
ORZHVTYKPFFVMG-UHFFFAOYSA-N
2-[({5-[3-(3-{[bis(carboxymethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxymethyl)amino]acetic acid
CC1=CC(=CC(CN(CC(O)=O)CC(O)=O)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC(O)=O)CC(O)=O)=C1
This item is not returnable. View return policy
1611-35-4
C31H32N2O13S
672.66
11493
ORZHVTYKPFFVMG-UHFFFAOYSA-N
2-[({5-[3-(3-{[bis(carboxymethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxymethyl)amino]acetic acid
CC1=CC(=CC(CN(CC(O)=O)CC(O)=O)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC(O)=O)CC(O)=O)=C1
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'