missing translation for 'onlineSavingsMsg'
Learn More

o-Dianisidine dihydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11403397
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is suitable for use in ELISA procedures. In neurohistochemistry o-Dianisidine dihydrochloride has demonstrated perikaryal labeling as well as retrograde and anterograde transport to the primary olfactory cortex. It has also been used for a paper based-micro fluidic device to semi-quantitatively measure lactate, uric acid, and glucose. 3,3'-Dimethoxybenzidine is used principally as an intermediate in the production of commercial bisazobiphenyl dyes for coloring textiles, paper, plastic, rubber, and leather.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is suitable for use in ELISA procedures. In neurohistochemistry o-Dianisidine dihydrochloride has demonstrated perikaryal labeling as well as retrograde and anterograde transport to the primary olfactory cortex. It has also been used for a paper based-micro fluidic device to semi-quantitatively measure lactate, uric acid, and glucose. 3,3′-Dimethoxybenzidine is used principally as an intermediate in the production of commercial bisazobiphenyl dyes for coloring textiles, paper, plastic, rubber, and leather.

Solubility
Soluble in water (50 mg/ml). It dissolves in ethyl alcohol ,acid and in organic solvent.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20325-40-0
Molecular Formula C14H18Cl2N2O2
Molecular Weight (g/mol) 317.21
MDL Number MFCD00012488
InChI Key UXTIAFYTYOEQHV-UHFFFAOYSA-N
Synonym 3,3'-dimethoxybenzidine dihydrochloride, o-dianisidine dihydrochloride, 3,3'-dimethoxy-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-qv96qa6ukk, dianisidine dihydrochloride, ccris 998, o-dianisidine hcl, qv96qa6ukk, c.i. disperse black 6 dihydrochloride
PubChem CID 62311
IUPAC Name 4-(4-amino-3-methoxyphenyl)-2-methoxyaniline;dihydrochloride
SMILES COC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)OC)N.Cl.Cl

Specifications

Melting Point 268°C (decomposition)
Assay Percent Range 99%
Quantity 1 g
Beilstein 3917996
Solubility Information Soluble in water (50mg/ml). It dissolves in ethyl alcohol ,acid and in organic solvent.
Formula Weight 317.21
Percent Purity 99%
Chemical Name or Material o-Dianisidine dihydrochloride
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H350-May cause cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 3,3-Dimethoxybenzidine dihydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
o-Dianisidine dihydrochloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.