missing translation for 'onlineSavingsMsg'
Learn More

O-Methyl-L-tyrosine, 98%, Thermo Scientific Chemicals

Product Code. 15459598
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

O-Methyl-L-tyrosine, is used as a fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-Methyl-L-tyrosine, is used as a fine chemical intermediate.

Solubility
Very soluble in water.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6230-11-1
MDL Number MFCD00002604
ChEBI CHEBI:29003

Specifications

Melting Point 259°C to 261°C (decomposition)
Molecular Formula C5H8O5
Beilstein 2212726
Synonym s-2-hydroxy-2-methylsuccinic acid, l-citramalic acid, s-+-citramalic acid, s-citramalic acid, s-2-methylmalic acid, unii-b3fc6pa80i, b3fc6pa80i, 2s-2-hydroxy-2-methylbutanedioic acid, l-alpha-hydroxypyrotartaric acid, 2s-2-hydroxy-2-methylsuccinic acid
Solubility Information Very soluble in water.
InChI Key XFTRTWQBIOMVPK-YFKPBYRVSA-N
SMILES CC(CC(=O)O)(C(=O)O)O
IUPAC Name (2S)-2-hydroxy-2-methylbutanedioic acid
Molecular Weight (g/mol) 148.11
PubChem CID 441696
Formula Weight 195.22
Percent Purity 98%
Quantity 1 g
Physical Form Solid
Chemical Name or Material O-Methyl-L-tyrosine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'