missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals O-Methyl-L-tyrosine, 98%

Product Code. 10042150 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10042150

Brand: Thermo Scientific Chemicals 169750010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6230-11-1 | C10H13NO3 | 195.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6230-11-1
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.22
MDL Number MFCD00002604
InChI Key GEYBMYRBIABFTA-IMWMWJONNA-N
Synonym 4-methoxy-l-phenylalanine, o-methyl-l-tyrosine, h-tyr me-oh, o-methyltyrosine, s-2-amino-3-4-methoxyphenyl propanoic acid, p-methoxy-l-phenylalanine, 2s-2-amino-3-4-methoxyphenyl propanoic acid, tyrosine, o-methyl, l-4-methoxyphenylalanine, h-4-methoxy-phe-oh
PubChem CID 2723935
IUPAC Name (2S)-2-amino-3-(4-methoxyphenyl)propanoic acid
SMILES COC1=CC=C(C[C@H](N)C(O)=O)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula CH3OC6H4CH2CH(NH2)CO2H
Beilstein 14, I, 668
Specific Rotation -7° to -9° (20°C, 589nm) (c=2, 1N HCl)
Specific Rotation Condition −8° (20°C c=2,1N HCl)
Quantity 1 g
Formula Weight 195.22
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material O-Methyl-L-tyrosine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.