missing translation for 'onlineSavingsMsg'
Learn More

o-Nitroacetophenone, 95%, Thermo Scientific Chemicals

£25.70 - £96.70

Chemical Identifiers

CAS 577-59-3
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.15
MDL Number MFCD00007144
InChI Key SUGXZLKUDLDTKX-UHFFFAOYSA-N
Synonym 2'-nitroacetophenone, 1-2-nitrophenyl ethanone, o-nitroacetophenone, ethanone, 1-2-nitrophenyl, 2-acetylnitrobenzene, acetophenone, 2'-nitro, methyl 2-nitrophenyl ketone, unii-52l53k4x3o, ccris 2329, 1-2-nitrophenyl ethan-1-one
PubChem CID 11346
IUPAC Name 1-(2-nitrophenyl)ethanone
SMILES CC(=O)C1=CC=CC=C1[N+]([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10113230
View Documents
Thermo Scientific Acros
128320025
2.5 g Glass bottle
£25.70
2.50g
Please to purchase this item. Need a web account? Register with us today!
10755231
View Documents
Thermo Scientific Acros
128320200
20 g Glass bottle
£96.70
20g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

577-59-3
165.15
SUGXZLKUDLDTKX-UHFFFAOYSA-N
11346
CC(=O)C1=CC=CC=C1[N+]([O-])=O
C8H7NO3
MFCD00007144
2'-nitroacetophenone, 1-2-nitrophenyl ethanone, o-nitroacetophenone, ethanone, 1-2-nitrophenyl, 2-acetylnitrobenzene, acetophenone, 2'-nitro, methyl 2-nitrophenyl ketone, unii-52l53k4x3o, ccris 2329, 1-2-nitrophenyl ethan-1-one
1-(2-nitrophenyl)ethanone

Specifications

577-59-3
100.0
159.0°C (16.0 mmHg)
94% min. (GC)
O2NC6H4COCH3
MFCD00007144
2'-nitroacetophenone, 1-2-nitrophenyl ethanone, o-nitroacetophenone, ethanone, 1-2-nitrophenyl, 2-acetylnitrobenzene, acetophenone, 2'-nitro, methyl 2-nitrophenyl ketone, unii-52l53k4x3o, ccris 2329, 1-2-nitrophenyl ethan-1-one
SUGXZLKUDLDTKX-UHFFFAOYSA-N
1-(2-nitrophenyl)ethanone
11346
95%
94.0
28.0°C to 30.0°C
>112°C
C8H7NO3
1.5500 to 1.5520
07, 288
Solubility in water: insoluble. Other solubilities: soluble in ethanol and ether
CC(=O)C1=CC=CC=C1[N+]([O-])=O
165.15
165.15
o-Nitroacetophenone, 95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 209-414-8

missing translation for 'rtecsNumber' : AM9600000

missing translation for 'tsca' : TSCA

Product Identifier
  • o-Nitroacetophenone

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only