missing translation for 'onlineSavingsMsg'
Learn More

o-Phthaloyl dichloride, 90%, remainder mainly phthalic acid, Thermo Scientific Chemicals

£17.19 - £94.76

Chemical Identifiers

CAS 88-95-9
MDL Number MFCD00000666
InChI Key FYXKZNLBZKRYSS-UHFFFAOYSA-N
Synonym phthaloyl chloride, phthaloyl dichloride, phthalic chloride, phthalyl chloride, phthalic dichloride, phthalic acid dichloride, phthalyl dichloride, 1,2-benzenedicarbonyl dichloride, o-phthaloyl dichloride, phthaloyldichloride
PubChem CID 6955
IUPAC Name benzene-1,2-dicarbonyl chloride
SMILES C1=CC=C(C(=C1)C(=O)Cl)C(=O)Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10745611
View Documents
Thermo Scientific Acros
131110050
5 mL Glass bottle
£17.19
5mL
Please to purchase this item. Need a web account? Register with us today!
10031400
View Documents
Thermo Scientific Acros
131111000
100 mL Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10157753
View Documents
Thermo Scientific Acros
131115000
500 mL Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

88-95-9
FYXKZNLBZKRYSS-UHFFFAOYSA-N
6955
C1=CC=C(C(=C1)C(=O)Cl)C(=O)Cl
MFCD00000666
phthaloyl chloride, phthaloyl dichloride, phthalic chloride, phthalyl chloride, phthalic dichloride, phthalic acid dichloride, phthalyl dichloride, 1,2-benzenedicarbonyl dichloride, o-phthaloyl dichloride, phthaloyldichloride
benzene-1,2-dicarbonyl chloride

Specifications

88-95-9
Yellow
269.0°C to 271.0°C
Authentic
C8H4Cl2O2
1.5674 to 1.5694
09, 805
15, 7487
phthaloyl chloride, phthaloyl dichloride, phthalic chloride, phthalyl chloride, phthalic dichloride, phthalic acid dichloride, phthalyl dichloride, 1,2-benzenedicarbonyl dichloride, o-phthaloyl dichloride, phthaloyldichloride
FYXKZNLBZKRYSS-UHFFFAOYSA-N
benzene-1,2-dicarbonyl chloride
6955
90%
o-Phthaloyl dichloride, Remainder mainly phthalic acid
12.0°C
1.4000g/mL
113°C
90%
C6H4(COCl)2
MFCD00000666
01,882; 14,263
1.4
Solubility in water: decomposes. Other solubilities: soluble in ether, decomposes in alcohol
C1=CC=C(C(=C1)C(=O)Cl)C(=O)Cl
203.02
203.02
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful in contact with skin.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory ir

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 201-869-

Product Identifier
  • o-Phthaloyl dichloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : 1,2-Benzenedicarbonyl dichloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only