Learn More
Cytiva O-ring
Use in aqueous solutions and nearly all organic solvents commonly used in liquid chromatography of macromolecules. Cytiva Life Sciences™ O-rings are made of nitrile rubber.
£22.00 - £198.00
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
11785185
|
Cytiva
18-1104-97 |
11.3 x 2.4mm |
£198.00
Each |
In Stock
Log in to see stock. |
|||||
10201640
|
Cytiva
19-0685-01 |
37.47 x 5.34mm |
£79.00
Pack of 5 |
Estimated Shipment: 14-02-2025
Log in to see stock. |
|||||
11570854
|
Cytiva
19-7595-01 |
27.1 x 1.60 mm |
£22.00
Pack of 5 |
In Stock
Log in to see stock. |
|||||
11530854
|
Cytiva
19-5043-01 |
6.75 x 1.78mm |
£50.00
Pack of 10 |
In Stock
Log in to see stock. |
|||||
Description
- Can be used in aqueous solutions
- Nearly all organic solvents
- The following few exceptions: chlorinated hydrocarbons, acetone and otherketones, aliphatic esters and phenol. Solutions containingmore than 10% NaOH, 10% HCl, other strong mineral acidsor 5% acetic acid should not be used
- Made of Nitrile Rubber (NBR)
Specifications
O-Ring |