Learn More
RETSCH O-Ring for Test Sieves
Placed in the test sieve's recess to provide maximum stability and optimum sealing when used in sieve stacks. These o-rings for test sieves are available in several sizes, based upon the test sieve's diameter.
Brand: RETSCH 05.114.0045
Code : NEW
Additional Details : Weight : 0.00200kg
Description
- Provides maximum stability and optimum sealing when used in sieve stacks
- Available in several sizes, based upon the test sieve's diameter
Specifications
O-Ring | |
1Each | |
100 mm |