Learn More
RETSCH O-Ring for Test Sieves
Placed in the test sieve's recess to provide maximum stability and optimum sealing when used in sieve stacks. These o-rings for test sieves are available in several sizes, based upon the test sieve's diameter.
Brand:  RETSCH 05.114.0048
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 11944432
  • £8.00 / Each
Estimated Shipment: 07-10-2024
to see stock.
Add to basket
Description
Description
  • Provides maximum stability and optimum sealing when used in sieve stacks
  • Available in several sizes, based upon the test sieve's diameter
Specifications
Specifications
O-Ring
1Each
400 mm