Learn More
DWK Life Sciences O-ring, for stopcock keys with groove
Shop All DWK Life Sciences Products
Click to view available options
Description:
10.0 NS
21.5 NS
24.0 NS
29.2 NS
Unit Size:
Pack of 50
Description
The O-ring in the groove of the DURAN™ tap plug provided for this purpose secures the tap plug in the disposable tap.
- Secures the valve plug from falling out of the valve
Specifications
Specifications
| For Use With (Application) | Sealing |
| Quantity | 50/Pk. |
| Description | 10.0 NS |