Learn More
DWK Life Sciences O-Ring, transparent, from silicone (VMQ), not suitable for desiccators
Seal is suitable for the tight sealing of flat flange vessels with annular groove.
£46.80 - £191.00
Specifications
Material Silicone (VMQ)
Quantity 5/Pk.
For Use With (Application) Tight closure of flange vessels
View More Specs
Products 5
Product Code Brand Diameter (Metric) Outside Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outside Price Quantity & Availability  
11890680
View Documents
DWK Life Sciences
292256106
215 mm
£191.00
Pack of 5
Estimated Shipment: 15-01-2025
to see stock.
Add to basket
15282119
View Documents
DWK Life Sciences
292254602
110 mm
£73.55
Pack of 5
In Stock
to see stock.
Add to basket
13497068
View Documents
DWK Life Sciences
292255101
133 mm
£112.00
Pack of 5
In Stock
to see stock.
Add to basket
11880680
View Documents
DWK Life Sciences
292255701
157 mm
£150.00
Pack of 5
In Stock
to see stock.
Add to basket
11870680
View Documents
DWK Life Sciences
292253409
75 mm
£46.80
Pack of 5
In Stock
to see stock.
Add to basket
Description
Description
Flat flange vessels are large-volume reaction vessels consisting of a lower part and a lid. These two parts are held together by a clamping ring. The transparent O-ring made of silicone serves as a seal for flat flange vessels with an annular groove. It prevents tilting of the ground surfaces between the lower part and the lid and convinces by its good elasticity. The chemical resistance is limited compared to FEP coated O-rings.
  • Accessories for flat flange vessels from silicone (VMQ), so is highly elastic
  • The chemical resistance of silicone is lower than FEP coated O-rings
Specifications
Specifications
Silicone (VMQ)
5/Pk.
Tight closure of flange vessels