missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-tert-Butyl-D-serine methyl ester hydrochloride, 98%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Color | White |
Beilstein | 2041979 |
Formula Weight | 211.69 |
Percent Purity | 98% |
Quantity | 5 g |
Physical Form | Powder |
Chemical Name or Material | O-tert-Butyl-D-serine methyl ester hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'