missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-tert-Butyl-L-serine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
O-tert-Butyl-L-serine tert-butyl ester hydrochloride is a protected L-serine used in peptide synthesis, e.g. synthesis of glycopeptides and lipopeptides and preparation of amino-phospholipids. O-tert-Butyl-L-serine (Ser(TBU-OH), a serine hydroxyl protected amino acid, is used in SPPS such as the synthesis of tailed cyclic RGD peptides to prevent glutarimide formation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
O-tert-Butyl-L-serine tert-butyl ester hydrochloride is a protected L-serine used in peptide synthesis, e.g. synthesis of glycopeptides and lipopeptides and preparation of amino-phospholipids. O-tert-Butyl-L-serine (Ser(TBU-OH), a serine hydroxyl protected amino acid, is used in SPPS such as the synthesis of tailed cyclic RGD peptides to prevent glutarimide formation.
Solubility
Soluble in water.
Notes
Light Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, heat.
O-tert-Butyl-L-serine tert-butyl ester hydrochloride is a protected L-serine used in peptide synthesis, e.g. synthesis of glycopeptides and lipopeptides and preparation of amino-phospholipids. O-tert-Butyl-L-serine (Ser(TBU-OH), a serine hydroxyl protected amino acid, is used in SPPS such as the synthesis of tailed cyclic RGD peptides to prevent glutarimide formation.
Solubility
Soluble in water.
Notes
Light Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, heat.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 220°C (decomposition) |
Color | White |
Beilstein | 2500118 |
Solubility Information | Soluble in water. |
Formula Weight | 161.2 |
Percent Purity | 98% |
Quantity | 1 g |
Physical Form | Powder |
Chemical Name or Material | O-tert-Butyl-L-serine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'